Bis(3-fluorphenyl)disulfid ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B3044-1G B3044-5G
TCIAB3044-1GEA 304 DKK
TCIAB3044-1G TCIAB3044-5G
Bis(3-fluorphenyl)disulfid ≥97.0% (by GC)
Bis(3-fluorphenyl)disulfid
Formel: C₁₂H₈F₂S₂
MDL Number: MFCD01318104
CAS-nummer: 63930-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=474be3ce71d658dff7ac29156737deb914ec767b&vid=474be3ce71d658dff7ac29156737deb914ec767b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR