Ethyl 6-Bromhexanoat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
6-Bromocaproic acid ethyl ester, 6-Bromohexanoic acid ethyl ester, Ethyl 6-bromohexanoate
B1465-25G
B1465-250G
TCIAB1465-25GEA
403
DKK
TCIAB1465-25G
TCIAB1465-250G
Ethyl 6-Bromhexanoat ≥98.0% (by GC)
Ethyl 6-Bromhexanoat
Formel:
BrH₂C(CH₂)₄COOCH₂CH₃ Kogepunkt: 130°C Smeltepunkt: 33 °C Massefylde: 1,2592 g/cm³ (20 °C) Flammepunkt: 58°C |
MDL Number:
MFCD00000270 CAS-nummer: 25542-62-5 EINECS: 247-085-2 UN nummer: 3272 ADR: 3,III |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...