α-Angelicalacton ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A1090-25G
A1090-100G
TCIAA1090-25GEA
419
DKK
TCIAA1090-25G
TCIAA1090-100G
α-Angelicalacton ≥98.0% (by GC)
α-Angelicalacton
Formel:
C₅H₆O₂ Kogepunkt: 56°C Smeltepunkt: 0 °C Massefylde: 1,0904 g/cm³ (12 °C) Flammepunkt: 68°C |
MDL Number:
MFCD00005375 CAS-nummer: 591-12-8 EINECS: 209-701-8 Merck Index: 14,00647 |
|
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |