5-fluorcytosin ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one
F0321-1G
F0321-25G
F0321-5G
TCIAF0321-1GEA
206
DKK
TCIAF0321-1G
TCIAF0321-25G
TCIAF0321-5G
5-fluorcytosin ≥98.0% (by HPLC, titration analysis)
5-fluorcytosin
Formel:
C₄H₄FN₃O Molekylvægt: 129.09 g/mol Kogepunkt: 298 Smeltepunkt: 165 |
MDL Number:
MFCD00006035 CAS-nummer: 2022-85-7 EINECS: 217-968-7 Merck Index: 14,04125 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...