2-Acetylcyclopentanon ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A0869-25ML A0869-5ML
TCIAA0869-25MLEA 1820 DKK
TCIAA0869-25ML TCIAA0869-5ML
2-Acetylcyclopentanon ≥95.0% (by GC)
2-Acetylcyclopentanon
Formel: C₇H₁₀O₂
Kogepunkt: 103°C
Massefylde: 1,06 g/cm³ (20 °C)
Flammepunkt: 72°C
MDL Number: MFCD00001413
CAS-nummer: 1670-46-8
EINECS: 216-797-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55efbd46bd0db09e217494c9a162772df1fb752f&vid=55efbd46bd0db09e217494c9a162772df1fb752f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR