5-Acetyl-2,4-dimethylthiazol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A1215-10G
A1215-25G
TCIAA1215-10GEA
206
DKK
TCIAA1215-10G
TCIAA1215-25G
5-Acetyl-2,4-dimethylthiazol ≥98.0% (by GC)
5-Acetyl-2,4-dimethylthiazol
Formel:
C₇H₉NOS Kogepunkt: 230°C Massefylde: 1,15 g/cm³ (20 °C) Flammepunkt: 104°C |
MDL Number:
MFCD00005331 CAS-nummer: 38205-60-6 EINECS: 253-826-0 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...