4-Brom-3,5-xylenol ≥98.0% (by GC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 4-Bromo-3,5-dimethylphenol, 2-Bromo-5-hydroxy-3-xylene
B2146-25G B2146-5G
TCIAB2146-25GEA 649 DKK
TCIAB2146-25G TCIAB2146-5G
4-Brom-3,5-xylenol ≥98.0% (by GC)
4-Brom-3,5-xylenol
Formel: C₈H₉BrO
Molekylvægt: 201.06 g/mol
Kogepunkt: 259
Smeltepunkt: 116
MDL Number: MFCD00002315
CAS-nummer: 7463-51-6
EINECS: 231-255-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcf7d6890416da34540ddb638f80edf57e5d7e2b&vid=fcf7d6890416da34540ddb638f80edf57e5d7e2b&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 115.0 to 118.0 deg-C
Solubility in Methanol very faint turbidity