5-Amino-o-cresol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 5-Amino-o-cresol, 3-Hydroxy-4-methylaniline, 1-Amino-3-hydroxy-4-methylbenzene, 5-Amino-2-methylphenol, 4-Amino-2-hydroxytoluene
A0925-25G A0925-500G
TCIAA0925-25GEA 353 DKK
TCIAA0925-25G TCIAA0925-500G
5-Amino-o-cresol ≥98.0% (by GC, titration analysis)
5-Amino-o-cresol
Formel: C₇H₉NO
Molekylvægt: 123.15 g/mol
Kogepunkt: 259
Smeltepunkt: 162
MDL Number: MFCD00043922
CAS-nummer: 2835-95-2
EINECS: 220-618-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af4a5e5a8a2ca38132d65dd5d7aa27bba4bfd1d0&vid=af4a5e5a8a2ca38132d65dd5d7aa27bba4bfd1d0&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Light yellow to Dark green powder to crystaline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 160.0 to 163.0 deg-C