N,N-Diethyl-1,3-diaminopropan ≥99.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
3-(Diethylamino)propylamin
D0473-25ML
D0473-500ML
TCIAD0473-25MLEA
132
DKK
TCIAD0473-25ML
TCIAD0473-500ML
N,N-Diethyl-1,3-diaminopropan ≥99.0% (by GC, titration analysis)
N,N-Diethyl-1,3-diaminopropan
Formel:
C₇H₁₈N₂ Molekylvægt: 130.23 g/mol Kogepunkt: 171°C Smeltepunkt: -60 Flammepunkt: 59°C |
MDL Number:
MFCD00008218 CAS-nummer: 104-78-9 EINECS: 203-236-4 UN nummer: 2684 ADR: 3 III |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...