2-Amino-p-cresol ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: 3-Amino-4-hydroxytoluene, 1-Amino-2-hydroxy-5-methylbenzene, 2-Amino-4-methylphenol, 2-Hydroxy-5-methylaniline
A0330-25G A0330-500G A0330-100G
TCIAA0330-25GEA 345 DKK
TCIAA0330-25G TCIAA0330-500G TCIAA0330-100G
2-Amino-p-cresol ≥98.0% (by GC, titration analysis)
2-Amino-p-cresol
Formel: C₇H₉NO
Molekylvægt: 123.15 g/mol
Kogepunkt: 259
MDL Number: MFCD00007699
CAS-nummer: 95-84-1
EINECS: 202-457-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79124c1e313f254c348db95b8fe5ee8101e7037f&vid=79124c1e313f254c348db95b8fe5ee8101e7037f&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Light red to Green powder to crystalline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 134.0 to 139.0 deg-C