2,6-Dichlor-p-cresol ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,6-Dichloro-4-methylphenol

D2173-1G D2173-5G
TCIAD2173-1GEA 222 DKK
TCIAD2173-1G TCIAD2173-5G
2,6-Dichlor-p-cresol ≥96.0% (by GC)
2,6-Dichlor-p-cresol
Formel: C₇H₆Cl₂O
Molekylvægt: 177.03 g/mol
MDL Number: MFCD00134679
CAS-nummer: 2432-12-4
EINECS: 219-402-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9af66cd46dd240f2d96169e7b773207b4324a23&vid=c9af66cd46dd240f2d96169e7b773207b4324a23&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR