3,5-Dibenzyloxybenzylalkohol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (3,5-Bis(benzyloxy)phenyl)methanol, 3,5-Bis(benzyloxy)benzyl alcohol

D2651-25G D2651-5G
TCIAD2651-25GEA 2160 DKK
TCIAD2651-25G TCIAD2651-5G
3,5-Dibenzyloxybenzylalkohol ≥98.0% (by GC)
3,5-Dibenzyloxybenzylalkohol
Formel: C₂₁H₂₀O₃
Smeltepunkt: 85 °C
MDL Number: MFCD01863236
CAS-nummer: 24131-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51c83db5db9d6e2e26228e58a9b0c6e81d392833&vid=51c83db5db9d6e2e26228e58a9b0c6e81d392833&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR