trans-Kanelaldehyd ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C0352-25ML C0352-500ML
TCIAC0352-25MLEA 132 DKK
TCIAC0352-25ML TCIAC0352-500ML
trans-Kanelaldehyd ≥98.0% (by GC)
trans-Kanelaldehyd
Formel: C₉H₈O
Kogepunkt: 252 °C (1013 hPa)
Smeltepunkt: -8°C
Massefylde: 1,05 g/cm³ (20 °C)
Flammepunkt: 129°C
MDL Number: MFCD00007000
CAS-nummer: 14371-10-9
Merck Index: 14,02297
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5239c6e52473c74871574276574883fc3da4213&vid=c5239c6e52473c74871574276574883fc3da4213&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR