Crotonsyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: trans-Crotonic acid, 2-Butenoic acid, trans-2-Butenoic acid

C0416-25G C0416-100G
TCIAC0416-25GEA 181 DKK
TCIAC0416-25G TCIAC0416-100G
Crotonsyre ≥98.0% (by GC, titration analysis)
Crotonsyre
Formel: C₄H₆O₂
Kogepunkt: 185°C
Smeltepunkt: 71…72 °C
Massefylde: 1,02 g/cm³ (20 °C)
Flammepunkt: 90 °C
MDL Number: MFCD00002701
CAS-nummer: 107-93-7
EINECS: 203-533-9
UN nummer: 2823
ADR: 8,III
Merck Index: 14,02597
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=467683da81015d8f2dfcbb88fb8aae740abe94c1&vid=467683da81015d8f2dfcbb88fb8aae740abe94c1&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 71.0 to 73.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR