Malononitril ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

M0033-25G M0033-500G M0033-100G
TCIAM0033-25GEA 132 DKK
TCIAM0033-25G TCIAM0033-500G TCIAM0033-100G
Malononitril ≥98.0% (by GC, titration analysis)
Malononitril
Formel: C₃H₂N₂
Kogepunkt: 220 °C (1013 hPa)
Smeltepunkt: 30…32 °C
Massefylde: 1,191 g/cm³ (20 °C)
Flammepunkt: 112 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00001883
CAS-nummer: 109-77-3
EINECS: 203-703-2
UN nummer: 2647
ADR: 6.1,II
Merck Index: 14,05711
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c5186e4b34eadbcdb8884da1e9e36931b9b0058&vid=9c5186e4b34eadbcdb8884da1e9e36931b9b0058&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR