tert-Butylbromacetat ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Bromoacetic tert-butyl ester

B1473-25G B1473-250G
TCIAB1473-25GEA 230 DKK
TCIAB1473-25G TCIAB1473-250G
tert-Butylbromacetat ≥98.0% (by GC)
tert-Butylbromacetat
Formel: BrCH₂COOC(CH₃)₂
Kogepunkt: 76°C
Massefylde: 1,338 g/cm³ (20 °C)
Flammepunkt: 49°C
MDL Number: MFCD00000188
CAS-nummer: 5292-43-3
EINECS: 226-133-6
UN nummer: 2924
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f4d7c239c54edfaeb33138007fbb73ab7f1ebec&vid=1f4d7c239c54edfaeb33138007fbb73ab7f1ebec&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR