5-Methoxy-2-methylindol ≥99.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5-Methoxy-2-methyl-1H-indole

M0730-1G M0730-5G
TCIAM0730-1GEA 854 DKK
TCIAM0730-1G TCIAM0730-5G
5-Methoxy-2-methylindol ≥99.0% (by GC)
5-Methoxy-2-methylindole
Formel: C₁₀H₁₁NO
Molekylvægt: 161.2 g/mol
Kogepunkt: 145°C
Smeltepunkt: 89
MDL Number: MFCD00005620
CAS-nummer: 1076-74-0
EINECS: 214-066-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4115a018ed00745087213a4027c2631da1f93465&vid=4115a018ed00745087213a4027c2631da1f93465&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 86.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR