5-Chloro-2-thenoic acid ≥95.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

C1230-5G C1230-25G
TCIAC1230-5GEA 165 DKK
TCIAC1230-5G TCIAC1230-25G
5-Chloro-2-thenoic acid ≥95.0% (by GC, titration analysis)
5-Chloro-2-thenoic acid
Formel: C₅H₃ClO₂S
MDL Number: MFCD00041426
CAS-nummer: 24065-33-6
EINECS: 480-060-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75a5bd9eb9d9d49b62e32c3f0fbe07b060cd6406&vid=75a5bd9eb9d9d49b62e32c3f0fbe07b060cd6406&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR