5-(2-Bromphenyl)tetrazol ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-(2-Bromophenyl)tetrazole

B2577-25G B2577-5G
TCIAB2577-25GEA 1030 DKK
TCIAB2577-25G TCIAB2577-5G
5-(2-Bromphenyl)tetrazol ≥98.0% (by HPLC, titration analysis)
5-(2-Bromphenyl)tetrazol
Formel: C₇H₅BrN₄
Smeltepunkt: 183…184 °C
MDL Number: MFCD00151799
CAS-nummer: 73096-42-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80bb7015d0217a54ef84e086e69848fd6e2196ab&vid=80bb7015d0217a54ef84e086e69848fd6e2196ab&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 180.0 to 184.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR