Hexyl 4-hydroxybenzoat ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Hydroxybenzoic acid hexyl ester, Hexyl 4-hydroxybenzoate, Hexylparaben
H0213-25G
H0213-500G
H0213-100G
TCIAH0213-25GEA
419
DKK
TCIAH0213-25G
TCIAH0213-500G
TCIAH0213-100G
Hexyl 4-hydroxybenzoat ≥98.0% (by HPLC, titration analysis)
Hexyl 4-hydroxybenzoat
Formel:
C₁₃H₁₈O₃ Kogepunkt: 171°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00016480 CAS-nummer: 1083-27-8 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 50.0 to 55.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...