4-Brom-3,5-dimethylisoxazol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

B3519-1G B3519-5G
TCIAB3519-1GEA 230 DKK
TCIAB3519-1G TCIAB3519-5G
4-Brom-3,5-dimethylisoxazol ≥98.0% (by GC)
4-Bromo-3,5-dimethylisoxazole
Formel: C₅H₆BrNO
Molekylvægt: 176.01 g/mol
Kogepunkt: 75°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00068187
CAS-nummer: 10558-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96b286cec4d3faf439f676ad4e939b2c1e73e349&vid=96b286cec4d3faf439f676ad4e939b2c1e73e349&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR