4-Amino-2,6-dimethylpyrimidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A2091-25G A2091-5G
TCIAA2091-25GEA 2010 DKK
TCIAA2091-25G TCIAA2091-5G
4-Amino-2,6-dimethylpyrimidin ≥98.0% (by GC, titration analysis)
4-Amino-2,6-dimethylpyrimidine
Formel: C₆H₉N₃
Kogepunkt: 254°C
MDL Number: MFCD00006106
CAS-nummer: 461-98-3
EINECS: 207-320-1
Merck Index: 12,05338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f4473fc32857276f045cc5e4ece46ef26166d65&vid=1f4473fc32857276f045cc5e4ece46ef26166d65&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 183.0 to 187.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR