4,4-Dimethylcyclohexanon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4,4-Dimethylcyclohexan-1-one

D4149-1G D4149-5G
TCIAD4149-1GEA 370 DKK
TCIAD4149-1G TCIAD4149-5G
4,4-Dimethylcyclohexanon ≥98.0% (by GC)
4,4-Dimethylcyclohexanone
Formel: C₈H₁₄O
Kogepunkt: 73°C
MDL Number: MFCD00234965
CAS-nummer: 4255-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4b7618d21a5b27931f2cd6821259dab0ec69b88&vid=d4b7618d21a5b27931f2cd6821259dab0ec69b88&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR