Ethyl benzoylacetat ≥95.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Ethyl 3-oxo-3-phenylpropanoate
B0097-25G
B0097-500G
B0097-100G
TCIAB0097-25GEA
132
DKK
TCIAB0097-25G
TCIAB0097-500G
TCIAB0097-100G
Ethyl benzoylacetat ≥95.0% (by GC, titration analysis)
Ethyl benzoylacetat
Formel:
C₁₁H₁₂O₃ Kogepunkt: 270°C Smeltepunkt: < 0 °C Massefylde: 1,114 g/cm³ (20 °C) Flammepunkt: 141°C |
MDL Number:
MFCD00009196 CAS-nummer: 94-02-0 EINECS: 202-295-3 Merck Index: 14,03767 |
|
Specifikationer Testresultater
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...