5-Fluor-2-nitrobenzoesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

F0694-25G F0694-5G
TCIAF0694-25GEA 1780 DKK
TCIAF0694-25G TCIAF0694-5G
5-Fluor-2-nitrobenzoesyre ≥98.0% (by GC, titration analysis)
5-Fluor-2-nitrobenzoesyre
Formel: C₇H₄FNO₄
MDL Number: MFCD00055635
CAS-nummer: 320-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7bf36792394f34ca5d9c25aed3fd2a153e3395c6&vid=7bf36792394f34ca5d9c25aed3fd2a153e3395c6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR