3-Amino-2-methoxypyridin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A2403-25G A2403-5G
TCIAA2403-25GEA 1500 DKK
TCIAA2403-25G TCIAA2403-5G
3-Amino-2-methoxypyridin ≥98.0% (by GC, titration analysis)
3-Amino-2-methoxypyridine
Formel: C₆H₈N₂O
Kogepunkt: 118°C
MDL Number: MFCD00833386
CAS-nummer: 20265-38-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5c6a5b0c5cad018baab6899756738b7b8d9086f&vid=d5c6a5b0c5cad018baab6899756738b7b8d9086f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR