Galdesyre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid, 3α,7α,12α-Trihydroxycholan-24-oic acid, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,7α,12α-Trihydroxycholanic acid, 3α,7α,12α-Trihydroxy-5β-cholanic acid

C0324-25G C0324-500G
TCIAC0324-25GEA 452 DKK
TCIAC0324-25G TCIAC0324-500G
Galdesyre ≥98.0% (by GC, titration analysis)
Galdesyre
Formel: C₂₄H₄₀O₅
Kogepunkt: 545 °C (1013 hPa)
Smeltepunkt: 199 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003672
CAS-nummer: 81-25-4
EINECS: 201-337-8
Merck Index: 14,02203
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bad5260a9a73d0269c31cb56fbebea5ea0b7a3c&vid=6bad5260a9a73d0269c31cb56fbebea5ea0b7a3c&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +33.0 to +38.0 deg (C=2, EtOH)
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR