Methyl3,5-dihydroxybenzoat ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Methyl 3,5-dihydroxybenzoate, 3,5-Dihydroxybenzoic acid methyl ester
D2215-25G
D2215-250G
TCIAD2215-25GEA
378
DKK
TCIAD2215-25G
TCIAD2215-250G
Methyl3,5-dihydroxybenzoat ≥98.0% (by GC)
Methyl3,5-dihydroxybenzoat
Formel:
C₈H₈O₄ Molekylvægt: 168.15 g/mol Kogepunkt: 300 |
MDL Number:
MFCD00002289 CAS-nummer: 2150-44-9 EINECS: 218-426-2 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 165.0 to 168.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...