3,5-Dibrombenzaldehyd ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D2832-25G D2832-5G
TCIAD2832-25GEA 1660 DKK
TCIAD2832-25G TCIAD2832-5G
3,5-Dibrombenzaldehyd ≥97.0% (by GC)
3,5-Dibromobenzaldehyde
Formel: C₇H₄Br₂O
MDL Number: MFCD00156887
CAS-nummer: 56990-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2f019af7f45bfb77765335c60f412f8c16dca47&vid=c2f019af7f45bfb77765335c60f412f8c16dca47&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 89.0 to 93.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR