Benzothiazol-2-thiol ≥99.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT

M0247-25G M0247-500G
TCIAM0247-25GEA 165 DKK
TCIAM0247-25G TCIAM0247-500G
Benzothiazol-2-thiol ≥99.0% (by HPLC)
Benzothiazol-2-thiol
Formel: C₇H₅NS₂
Kogepunkt: 302 °C (1013 hPa)
Smeltepunkt: 177…182 °C
Massefylde: 1,42 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005781
CAS-nummer: 149-30-4
EINECS: 205-736-8
UN nummer: 3077
ADR: 9,III
Merck Index: 14,05868
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cc4f6b9b8585329256e9824df3020684633ade2&vid=5cc4f6b9b8585329256e9824df3020684633ade2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 180.0 to 183.0 deg-C
Solubility in hot Methanol transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR