2-Mercapto-5-methylbenzimidazol
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1,3-Dihydro-5-methyl-2H-benzimidazole-2-thione, 2-Mercapto-5-methyl-1H-benzimidazole
M1357-100G
M1357-25G
TCIAM1357-100GEA
911
DKK
TCIAM1357-100G
TCIAM1357-25G
2-Mercapto-5-methylbenzimidazol
2-Mercapto-5-methylbenzimidazole
Formel:
C₈H₈N₂S Kogepunkt: 349 °C (1013 hPa) Smeltepunkt: 220 °C |
MDL Number:
MFCD00010617 CAS-nummer: 27231-36-3 EINECS: 248-350-5 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...