Isobutyronitril ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Isopropyl cyanide

I0112-25ML I0112-500ML
TCIAI0112-25MLEA 132 DKK
TCIAI0112-25ML TCIAI0112-500ML
Isobutyronitril ≥98.0% (by GC)
Isobutyronitril
Formel: C₄H₇N
Molekylvægt: 69.11 g/mol
Kogepunkt: 108°C
Smeltepunkt: –72°C
Flammepunkt: 8°C
MDL Number: MFCD00001873
CAS-nummer: 78-82-0
EINECS: 201-147-5
UN nummer: 2284
ADR: 3 II
Merck Index: 14,05156
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbe5e7dd74eb0a634d777e2b4f93d339e17cca1b&vid=fbe5e7dd74eb0a634d777e2b4f93d339e17cca1b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR