2-Chlor-6-trifluormethylpyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

C1986-1G C1986-5G
TCIAC1986-1GEA 304 DKK
TCIAC1986-1G TCIAC1986-5G
2-Chlor-6-trifluormethylpyridin ≥98.0% (by GC)
2-Chlor-6-trifluormethylpyridin
Formel: C₆H₃ClF₃N
Molekylvægt: 181.54 g/mol
Kogepunkt: 78°C
MDL Number: MFCD00042226
CAS-nummer: 39890-95-4
UN nummer: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd30824923bc7f941eda9a8cca1210a97852a428&vid=bd30824923bc7f941eda9a8cca1210a97852a428&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Freezing point 32.0 to 34.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR