2-Brom-6-(trifluormethyl)pyridin ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

B3759-1G B3759-5G
TCIAB3759-1GEA 222 DKK
TCIAB3759-1G TCIAB3759-5G
2-Brom-6-(trifluormethyl)pyridin ≥98.0% (by GC)
2-Bromo-6-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
Kogepunkt: 96°C
MDL Number: MFCD00153087
CAS-nummer: 189278-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ad6b2706e473eb31af1c2cceb90d7d2ad874531&vid=7ad6b2706e473eb31af1c2cceb90d7d2ad874531&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR