2-Aminobenzothiazol ≥97.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II
A0277-25G
A0277-500G
A0277-100G
TCIAA0277-25GEA
140
DKK
TCIAA0277-25G
TCIAA0277-500G
TCIAA0277-100G
2-Aminobenzothiazol ≥97.0% (by HPLC, titration analysis)
2-Aminobenzothiazole
Formel:
C₇H₆N₂S Molekylvægt: 150.2 g/mol |
MDL Number:
MFCD00005785 CAS-nummer: 136-95-8 EINECS: 205-268-4 Merck Index: 14,00424 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 126.0 to 131.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...