2-Amino-3-(trifluormethyl)pyridin 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
H34303.06
H34303.14
H34303.06EA
777
DKK
H34303.06
H34303.14
2-Amino-3-(trifluormethyl)pyridin 97%
2-Amino-3-(trifluoromethyl)pyridine
Formel:
C₆H₅F₃N₂ Smeltepunkt: 70…72 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04972538 CAS-nummer: 183610-70-0 UN nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...