1,3-Dimethyluracil ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1,3-Dimethylpyrimidine-2,4(1H,3H)-dione

D0808-1G D0808-25G D0808-5G
TCIAD0808-1GEA 312 DKK
TCIAD0808-1G TCIAD0808-25G TCIAD0808-5G
1,3-Dimethyluracil ≥98.0% (by GC)
1,3-Dimethyluracil
Formel: C₆H₈N₂O₂
Molekylvægt: 140.14 g/mol
MDL Number: MFCD00038065
CAS-nummer: 874-14-6
EINECS: 212-856-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71b5f0d43d54c4dc93ccbca639cb18620168e2ca&vid=71b5f0d43d54c4dc93ccbca639cb18620168e2ca&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 128.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR