Gemfibrozil ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: CI-719
G0368-25G G0368-5G
TCIAG0368-25GEA 1380 DKK
TCIAG0368-25G TCIAG0368-5G
Gemfibrozil ≥98.0% (by GC, titration analysis)
Gemfibrozil
Formel: C₁₅H₂₂O₃
Molekylvægt: 250.34 g/mol
Kogepunkt: 159°C
MDL Number: MFCD00079335
CAS-nummer: 25812-30-0
EINECS: 247-280-2
Merck Index: 14,04388
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff57660e610821b2f211872503b2b175f6d5499&vid=8ff57660e610821b2f211872503b2b175f6d5499&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 59.0 to 63.0 deg-C