(±)-Tetrahydrofurfurylalkohol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: (±)-2-Hydroxymethyloxolan

T0106-500ML T0106-25ML
TCIAT0106-500MLEA 238 DKK
TCIAT0106-500ML TCIAT0106-25ML
(±)-Tetrahydrofurfurylalkohol ≥98.0% (by GC)
(±)-Tetrahydrofurfurylalkohol
Formel: C₅H₁₀O₂
Kogepunkt: 176°C
Smeltepunkt: < –80 °C
Massefylde: 1,0543 g/cm³ (20 °C)
Flammepunkt: 75°C (closed cup)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005372
CAS-nummer: 97-99-4
EINECS: 202-625-6
Merck Index: 14,09213
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48f4c72bd2f5bbe843aad21a38f19c6943d52742&vid=48f4c72bd2f5bbe843aad21a38f19c6943d52742&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR