α,α,α,5-Tetrafluor-2-nitrotoluen ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-Fluoro-2-nitrobenzotrifluoride, α,α,α,5-Tetrafluoro-2-nitrotoluene, 1-Fluoro-4-nitro-3-(trifluoromethyl)benzene
F0497-25G
F0497-5G
TCIAF0497-25GEA
542
DKK
TCIAF0497-25G
TCIAF0497-5G
α,α,α,5-Tetrafluor-2-nitrotoluen ≥98.0% (by GC)
α,α,α,5-Tetrafluor-2-nitrotoluen
Formel:
C₇H₃F₄NO₂ Molekylvægt: 209.1 g/mol Kogepunkt: 199°C Smeltepunkt: 23.5 Massefylde: 1.49 |
MDL Number:
MFCD00039741 CAS-nummer: 393-09-9 |
|
Specifikationer Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...