p-Toluonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Methylbenzonitrile, p-Methylbenzonitrile
T0307-25G
T0307-500G
T0307-100G
TCIAT0307-25GEA
206
DKK
TCIAT0307-25G
TCIAT0307-500G
TCIAT0307-100G
p-Toluonitril ≥98.0% (by GC)
p-Toluonitril
Formel:
C₈H₇N Kogepunkt: 218°C Smeltepunkt: 25…28 °C Massefylde: 0,98 g/cm³ (20 °C) |
MDL Number:
MFCD00001827 CAS-nummer: 104-85-8 EINECS: 203-244-8 Merck Index: 14,09538 |
|
Specifikationer Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...