4,4'-Dichlorbenzhydrol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4,4'-Dichlorobenzhydryl alcohol, DBH, 4,4'-Dichlorophenyl phenyl carbinol, 4,4'-Dichloro-α-phenylbenzyl alcohol, 4,4'-Dichlorodiphenylmethanol, Bis(4-chlorophenyl)methanol, Bis(4-chlorophenyl)carbinol

D1915-25G D1915-250G
TCIAD1915-25GEA 329 DKK
TCIAD1915-25G TCIAD1915-250G
4,4'-Dichlorbenzhydrol ≥98.0% (by GC)
4,4'-Dichlorbenzhydrol
Formel: C₁₃H₁₀Cl₂O
MDL Number: MFCD00000629
CAS-nummer: 90-97-1
EINECS: 202-029-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ca624424b9daea1e5f3a5eca013a51b7167e32f&vid=9ca624424b9daea1e5f3a5eca013a51b7167e32f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 92.0 to 95.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR