3-Chlor-4-methylphenylisocyanat ≥97.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Chloro-4-isocyanato-1-methylbenzene
I0563-25G
I0563-5G
TCIAI0563-25GEA
903
DKK
TCIAI0563-25G
TCIAI0563-5G
3-Chlor-4-methylphenylisocyanat ≥97.0% (by GC)
3-Chlor-4-methylphenylisocyanat
Formel:
C₈H₆ClNO Kogepunkt: 124°C Massefylde: 1,224 g/cm³ (20 °C) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00013858 CAS-nummer: 28479-22-3 EINECS: 249-050-7 UN nummer: 3428 ADR: 6.1,II |
|
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange powder to lump |
Purity (GC) | min. 97.0 % |
Freezing point | 21.0 to 25.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...