4-Chlor-o-phenylendiamin ≥97.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Chloro-3,4-diaminobenzene, 4-Chlorobenzene-1,2-diamine, 4-Chloro-1,2-phenylenediamine, 4-Chloro-1,2-diaminobenzene
C0255-25G
C0255-5G
TCIAC0255-25GEA
386
DKK
TCIAC0255-25G
TCIAC0255-5G
4-Chlor-o-phenylendiamin ≥97.0% (by GC, titration analysis)
4-Chlor-o-phenylendiamin
Formel:
C₆H₇ClN₂ Kogepunkt: 280 °C (1013 hPa) Smeltepunkt: 89…90 °C |
MDL Number:
MFCD00011691 CAS-nummer: 95-83-0 EINECS: 202-456-8 |
|
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 72.0 to 76.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...