2-Chlorethyl methyl ether ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 2-Methoxyethyl chloride

C1173-25ML C1173-500ML C1173-100ML
TCIAC1173-25MLEA 173 DKK
TCIAC1173-25ML TCIAC1173-500ML TCIAC1173-100ML
2-Chlorethyl methyl ether ≥98.0% (by GC)
2-Chlorethyl methyl ether
Formel: C₃H₇ClO
Kogepunkt: 92°C
Smeltepunkt: –55…55 °C
Massefylde: 1,04 g/cm³ (20 °C)
Flammepunkt: 8°C
MDL Number: MFCD00000972
CAS-nummer: 627-42-9
EINECS: 210-999-7
UN nummer: 3271
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cd3ed0b93544bf2fddc8baaff46c9d791934be7&vid=9cd3ed0b93544bf2fddc8baaff46c9d791934be7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR