2-Chlorethyl ethyl ether ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Ethoxyethyl chloride, 2-Chlorodiethyl ether, 2-Chloroethylethylether
C1172-25ML
C1172-500ML
TCIAC1172-25MLEA
403
DKK
TCIAC1172-25ML
TCIAC1172-500ML
2-Chlorethyl ethyl ether ≥98.0% (by GC)
2-Chlorethyl ethyl ether
Formel:
C₄H₉ClO Kogepunkt: 108°C Smeltepunkt: –60 °C Massefylde: 1,0572 g/cm³ (0 °C) Flammepunkt: 6°C |
MDL Number:
MFCD00000974 CAS-nummer: 628-34-2 EINECS: 211-037-9 UN nummer: 3271 ADR: 3,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...