Bis(2-chlorethyl)ether ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Chloroethyl ether
B0472-25G
B0472-500G
TCIAB0472-25GEA
148
DKK
TCIAB0472-25G
TCIAB0472-500G
Bis(2-chlorethyl)ether ≥99.0% (by GC)
Bis(2-chlorethyl)ether
Formel:
C₄H₈Cl₂O Kogepunkt: 178°C Smeltepunkt: -47°C Massefylde: 1,22 g/cm³ (20 °C) Flammepunkt: 55°C |
MDL Number:
MFCD00000975 CAS-nummer: 111-44-4 EINECS: 203-870-1 UN nummer: 1916 ADR: 6.1,II Merck Index: 14,03066 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...