1-Butyl-3-methylimidazolium hexafluorphosphat ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Butyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V), 1-(But-1-yl)-3-methyl-1H-imidazol-3-ium hexafluorophosphate

B2320-25G B2320-5G
TCIAB2320-25GEA 632 DKK
TCIAB2320-25G TCIAB2320-5G
1-Butyl-3-methylimidazolium hexafluorphosphat ≥98.0% (by HPLC, total nitrogen)
1-Butyl-3-methylimidazolium hexafluorphosphat
Formel: C₈H₁₅F₆N₂P
Kogepunkt: ? 200 °C (1013 hPa)
Smeltepunkt: ∼6 °C
Massefylde: 1,38 g/cm³ (20 °C)
Flammepunkt: ≥ 110 °C
MDL Number: MFCD03093295
CAS-nummer: 174501-64-5
Merck Index: 14,01581
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a83f49974a470bb21566bd00833f3861823901ab&vid=a83f49974a470bb21566bd00833f3861823901ab&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR