1-Butyl-3-methylimidazolium chlorid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-Butyl-1-methyl-1H-imidazol-3-ium chloride

B2194-100G B2194-25G B2194-5G
TCIAB2194-100GEA 1220 DKK
TCIAB2194-100G TCIAB2194-25G TCIAB2194-5G
1-Butyl-3-methylimidazolium chlorid ≥98.0% (by HPLC, titration analysis)
1-Butyl-3-methylimidazolium chlorid
Formel: C₈H₁₅ClN₂
Molekylvægt: 174.67 g/mol
Smeltepunkt: 60
MDL Number: MFCD03095425
CAS-nummer: 79917-90-1
EINECS: 460-120-8
UN nummer: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3fee9e4cc7d9a12d5a4b03a1c69e225a2ae336a&vid=b3fee9e4cc7d9a12d5a4b03a1c69e225a2ae336a&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to lump
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR