p-Toluidiniumchlorid ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
p-Toluidine hydrochloride, 4-Toluidine HCl, p-Toluidine HCl, 4-Aminotoluene hydrochloride, 4-Methylaniline HCl, 4-Methylaniline hydrochloride, 4-Aminotoluene HCl, 4-Toluidine hydrochloride
T0303-25G
T0303-500G
TCIAT0303-25GEA
140
DKK
TCIAT0303-25G
TCIAT0303-500G
p-Toluidiniumchlorid ≥98.0% (by HPLC, total nitrogen)
p-Toluidiniumchlorid
Formel:
C₇H₁₀ClN Molekylvægt: 143.62 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00035497 CAS-nummer: 540-23-8 EINECS: 208-740-8 UN nummer: 2811 ADR: 6.1 III |
|
Specifikationer Testresultater
Appearance | White to Gray to Red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 241.0 to 245.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...